Bendamustine hydrochloride 99%
Nome del fornitore: Apollo Scientific
Vai a
Informazioni su questo articolo
Usage: Organic building block/reagent.
Sinonimi: Bendamustine HCl, 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochloride, SDX-15 HCl, Cytostasane HCl, EP 311, 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride
- Numero CAS:3543-75-7
- Formula:C₁₆H₂₂Cl₃N₃O₂
- Numero MDL:MFCD01658758
- Molecular Weight:394,72 g/mol
5 varianti
Product Details & Documents
Usage: Organic building block/reagent.
Sinonimi: Bendamustine HCl, 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochloride, SDX-15 HCl, Cytostasane HCl, EP 311, 4-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochloride
- Numero CAS:3543-75-7
- Formula:C₁₆H₂₂Cl₃N₃O₂
- Numero MDL:MFCD01658758
- Molecular Weight:394,72 g/mol
Product Family Options
Informazioni sul prodotto
- Tipo di conf.DisponibilitàPrice
- Specifiche:New ProductNew Product
- Specifiche:New ProductNew Product
- Specifiche:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



