Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Mandelic acid ≥95%

D-(-)-Mandelic acid ≥95%

New Product
Supplier:   Apollo Scientific
D-(-)-Mandelic acid ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
D-(-)-Mandelic acid ≥95%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
  • CAS Number:611-71-2
  • Formula:C₈H₈O₃
  • Molecular Weight:152,15 g/mol
Product Family Options

Product Information

  • Tipo di conf.
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 83.20
    SFr. 83.20
    1 * 5 g
       SFr. 83.20

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - SFr. 117.00
    SFr. 117.00
    1 * 10 g
       SFr. 117.00

    Specifications:
    New Product
    New Product