Allantoin ≥95%
Bottle
Codice catalogo APOSBUP19230-1ML
New Product
Nome del fornitore: Apollo Scientific
Allantoin ≥95%
Codice catalogo APOSBUP19230-1ML
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 97-59-6
Vai a
Product Details & Documents
Usage: Biochemical/life science applications.
Sinonimi: 1-(2,5-Dioxoimidazolidin-4-yl)urea
- Numero CAS:97-59-6
- Formula:C₄H₆N₄O₃
- Molecular Weight:158,12 g/mol
Specifiche
Specifiche
- Tipo di conf.:Bottle
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



