Angled Adapter, A854
Angle adapter for connecting wash items in load carriers with restricted loading height.
MIEL11035420
New Product
: MIELE
Suitable for connection to hoses with Luer-Lock
-
Also suitable, among other things, for connection to nozzles
Color: Stainless steel
Dimension: 24H×24W×33D
Weight: 0.01 kg
- Descrizione:Angle adapter for connecting wash items in load carriers with restricted loading height.