Orders LinkContactUsLinkComponent
MALDI Biotyper® Biotargets
MALDI Biotyper® Biotargets
Individually barcoded disposable 96 position MALDI target plates, Box of 20
Catalog # BRUK1840375
Supplier:   BRUKER DALTRONIK
MALDI Biotyper® Biotargets
Catalog # BRUK1840375
Supplier:  BRUKER DALTRONIK
CAS Number:  
MALDI Biotyper® Biotargets
Individually barcoded disposable 96 position MALDI target plates, Box of 20
CAS Number:  
Price
3 950.00 SFr.
Log in for availability
1 * 1 KIT
Product Details & Documents
The MBT Biotargets are equipped with hydrophobic coating on the target surrounding the hydrophilic sample positions facilitates the preparation of samples.
  • Disposable MALDI target plates
    96 sample positions
    Unique barcode for traceability
    Perfectly flat surface for reliable and reproducible results
    High flexibility in sample run sizes
    No wasted spots
    AnchorChip™ effect facilitates preparation
Documents
Specifications
Specifications
  • Descrizione:
    Individually barcoded disposable 96 position MALDI target plates
  • Pk Info:
    Box of 20
Specifications
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • BRUK8270006
    MBT Biotarget transport/storage boxes, 10