L-Carnosine
Nome del fornitore: G-Biosciences
Informazioni su questo articolo
L-Carnosine is a white to slightly off-white crystalline powder that is used as a substrate for carnosinase-1 and other enzymes.
Molecular Biology Grade
3 varianti
Product Details & Documents
L-Carnosine is a white to slightly off-white crystalline powder that is used as a substrate for carnosinase-1 and other enzymes.
Molecular Biology Grade
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- GENORC-679
- GENORC-677
- GENORC-681
- Protein/peptide name:
- L-Carnosine
- L-Carnosine
- L-Carnosine
- Protein synonyms:
- H-beta-Ala-His-OH| N-beta-Alanyl-L-histidine| beta-alanyl‑L-histidine
- CAS No.:
- 305-84-0
Specifiche
Product Family Options
Informazioni sul prodotto
- Tipo di conf.DisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.GENORC-679Protein/peptide nameL-CarnosineProtein synonymsCAS No. - Specifiche:
More
Specifications
Cat. No.GENORC-677Protein/peptide nameL-CarnosineProtein synonymsH-beta-Ala-His-OH| N-beta-Alanyl-L-histidine| beta-alanyl‑L-histidineCAS No.305-84-0 - Specifiche:
More
Specifications
Cat. No.GENORC-681Protein/peptide nameL-CarnosineProtein synonymsCAS No.