Software, SW8000
Communication software between the equipment and the PC, SW8000, Real-time display and recording, or post-cycle review
Codice catalogo ESPISW8000
Nome del fornitore: RAYPA
Vai a
Product Details & Documents
Allows the visualisation, documentation and printing of the processes carried out in the AE-B Series autoclaves.
- Includes 1,8 m Ethernet cable, USB stick with software, and USB-Ethernet adapter
- Real-time and post-cycle data analysis
- Displays batch data: Outcome, temperature, pressure, serial number, and more
- Encrypted cycle export (.DAT format)
- Password-protected access with multilingual interface (EN, ES, FR, IT)
Communication software between the equipment and the PC for display and recording in real time or display after each cycle. Cycles can also be printed or exported to excel. PC connection via ethernet. Data can also be exported directly to a USB memory stick.
Specifiche
- Descrizione:Communication software between the equipment and the PC, SW8000
- Per:Real-time display and recording, or post-cycle review