Orders LinkContactUsLinkComponent
VWR® Screw Cap Microtubes with Socket Caps
New Product
Supplier:   VWR Collection
VWR® Screw Cap Microtubes with Socket Caps
Supplier:  VWR Collection
New Product
Certificates
VWR® Screw Cap Microtubes with Socket Caps
VWR® Screw Cap Microtubes with Socket Caps
17 Options Available Below
Product Details & Documents
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
  • Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 and USP Class VI standards
  • High-clarity design: Crystal-clear walls for easy sample visibility
  • Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
  • Designed with an insertable ridge for convenient sample coding using colourful insert chips
  • Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
  • Sterile and non sterile options: Choose pre-sterilised assemblies or convenient non sterile combo packs
  • Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
    Maximum RCF: 20000×g
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
 : Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
 : Spare caps and colour insert chips also available.
Product Family Options
Screw Cap Microtubes and Socket Caps in Separate Resealable Bags
Product Information
  • Descrizione
    Capacità
    Colore
    Sterili
    Availability
    Price
  • Conical bottom microtubes with clear insertable flat cap, graduated, combo pack
    2,0 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 88.70
    SFr. 88.70
    1 * 500 ST
       SFr. 88.70
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear insertable flat cap, no graduations, combo pack
    0,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 85.80
    SFr. 85.80
    1 * 500 ST
       SFr. 85.80
    Specifications:
    New Product
    New Product
  • Conical bottom microtubes with clear insertable flat cap, graduated, combo pack
    1,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 88.70
    SFr. 88.70
    1 * 500 ST
       SFr. 88.70
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear insertable flat cap, graduated, combo pack
    2,0 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 83.00
    SFr. 83.00
    1 * 500 ST
       SFr. 83.00
    Specifications:
    New Product
    New Product
  • Conical bottom microtubes with clear insertable flat cap, no graduations, combo pack
    0,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 88.70
    SFr. 88.70
    1 * 500 ST
       SFr. 88.70
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear insertable flat cap, no graduations, combo pack
    1,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 85.80
    SFr. 85.80
    1 * 500 ST
       SFr. 85.80
    Specifications:
    New Product
    New Product
Sterile Assemblies: Screw Cap Microtubes with Attached Socket Caps
Product Information
  • Descrizione
    Capacità
    Colore
    Sterili
    Availability
    Price
  • Freestanding microtubes with clear insertable flat cap, no graduations, assembled pack
    1,5 ml
    trasparente
    +
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 138.00
    SFr. 138.00
    1 * 500 ST
       SFr. 138.00
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear insertable flat cap, graduated, assembled pack
    2,0 ml
    trasparente
    +
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 134.00
    SFr. 134.00
    1 * 500 ST
       SFr. 134.00
    Specifications:
    New Product
    New Product
  • Conical bottom microtubes with clear insertable flat cap, no graduations, assembled pack
    0,5 ml
    trasparente
    +
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 138.00
    SFr. 138.00
    1 * 500 ST
       SFr. 138.00
    Specifications:
    New Product
    New Product
  • Conical bottom microtubes with clear insertable flat cap, graduated, assembled pack
    2,0 ml
    trasparente
    +
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 138.00
    SFr. 138.00
    1 * 500 ST
       SFr. 138.00
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear insertable flat cap, no graduations, assembled pack
    0,5 ml
    trasparente
    +
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 138.00
    SFr. 138.00
    1 * 500 ST
       SFr. 138.00
    Specifications:
    New Product
    New Product
Screw Cap Microtubes with Tethered Socket Caps in Separate Resealable Bags
Product Information
  • Descrizione
    Capacità
    Colore
    Sterili
    Availability
    Price
  • Conical base microtubes with clear tethered, socket cap, no graduations, combo pack
    0,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 91.40
    SFr. 91.40
    1 * 500 ST
       SFr. 91.40
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack
    0,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 85.80
    SFr. 85.80
    1 * 500 ST
       SFr. 85.80
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack
    1,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 91.40
    SFr. 91.40
    1 * 500 ST
       SFr. 91.40
    Specifications:
    New Product
    New Product
  • Conical base microtubes with clear tethered, socket cap, graduated, combo pack
    2,0 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 91.40
    SFr. 91.40
    1 * 500 ST
       SFr. 91.40
    Specifications:
    New Product
    New Product
  • Conical base microtubes with clear tethered, socket cap, graduated, combo pack
    1,5 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 91.40
    SFr. 91.40
    1 * 500 ST
       SFr. 91.40
    Specifications:
    New Product
    New Product
  • Freestanding microtubes with clear tethered, socket cap, graduated, combo pack
    2,0 ml
    trasparente
    -
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 85.80
    SFr. 85.80
    1 * 500 ST
       SFr. 85.80
    Specifications:
    New Product
    New Product
Customisable Socket Caps
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Customisable screw caps, non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 57.00
    SFr. 57.00
    1 * 500 ST
       SFr. 57.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Customisable screw caps, non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 57.00
    SFr. 57.00
    1 * 500 ST
       SFr. 57.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Customisable screw caps, non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 54.80
    SFr. 54.80
    1 * 500 ST
       SFr. 54.80
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • Customisable screw caps, non sterile
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 57.00
    SFr. 57.00
    1 * 500 ST
       SFr. 57.00
Colour Inserts for Socket Caps
Product Information
  • Catalog #
    Descrizione
    Colore
    Availability
    Price
  • Socket cap insert, non sterile
    verde
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 30.10
    SFr. 30.10
    1 * 500 ST
       SFr. 30.10
Product Information
  • Catalog #
    Descrizione
    Colore
    Availability
    Price
  • Socket cap insert, non sterile
    blu
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 30.10
    SFr. 30.10
    1 * 500 ST
       SFr. 30.10
Product Information
  • Catalog #
    Descrizione
    Colore
    Availability
    Price
  • Socket cap insert, non sterile
    bianco
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 30.10
    SFr. 30.10
    1 * 500 ST
       SFr. 30.10