VWR® Screw Cap Microtubes with Socket Caps
About this item
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 and USP Class VI standards
- High-clarity design: Crystal-clear walls for easy sample visibility
- Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
- Designed with an insertable ridge for convenient sample coding using colourful insert chips
- Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
- Sterile and non sterile options: Choose pre-sterilised assemblies or convenient non sterile combo packs
-
Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
Maximum RCF: 20000×g
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
: Spare caps and colour insert chips also available.
17 Options Available Below
Product Details & Documents
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 and USP Class VI standards
- High-clarity design: Crystal-clear walls for easy sample visibility
- Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
- Designed with an insertable ridge for convenient sample coding using colourful insert chips
- Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
- Sterile and non sterile options: Choose pre-sterilised assemblies or convenient non sterile combo packs
-
Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
Maximum RCF: 20000×g
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
: Spare caps and colour insert chips also available.
Product Family Options
Screw Cap Microtubes and Socket Caps in Separate Resealable Bags
Product Information
- DescrizioneCapacitàColoreSteriliAvailabilityPrice
- Conical bottom microtubes with clear insertable flat cap, graduated, combo pack2,0 mltrasparente-Log in for availability1 * 500 STSFr. 88.70Specifications:New ProductNew Product
- Freestanding microtubes with clear insertable flat cap, no graduations, combo pack0,5 mltrasparente-Log in for availability1 * 500 STSFr. 85.80Specifications:New ProductNew Product
- Conical bottom microtubes with clear insertable flat cap, graduated, combo pack1,5 mltrasparente-Log in for availability1 * 500 STSFr. 88.70Specifications:New ProductNew Product
- Freestanding microtubes with clear insertable flat cap, graduated, combo pack2,0 mltrasparente-Log in for availability1 * 500 STSFr. 83.00Specifications:New ProductNew Product
- Conical bottom microtubes with clear insertable flat cap, no graduations, combo pack0,5 mltrasparente-Log in for availability1 * 500 STSFr. 88.70Specifications:New ProductNew Product
- Freestanding microtubes with clear insertable flat cap, no graduations, combo pack1,5 mltrasparente-Log in for availability1 * 500 STSFr. 85.80Specifications:New ProductNew Product
Sterile Assemblies: Screw Cap Microtubes with Attached Socket Caps
Product Information
- DescrizioneCapacitàColoreSteriliAvailabilityPrice
- Freestanding microtubes with clear insertable flat cap, no graduations, assembled pack1,5 mltrasparente+Log in for availability1 * 500 STSFr. 138.00Specifications:New ProductNew Product
- Freestanding microtubes with clear insertable flat cap, graduated, assembled pack2,0 mltrasparente+Log in for availability1 * 500 STSFr. 134.00Specifications:New ProductNew Product
- Conical bottom microtubes with clear insertable flat cap, no graduations, assembled pack0,5 mltrasparente+Log in for availability1 * 500 STSFr. 138.00Specifications:New ProductNew Product
- Conical bottom microtubes with clear insertable flat cap, graduated, assembled pack2,0 mltrasparente+Log in for availability1 * 500 STSFr. 138.00Specifications:New ProductNew Product
- Freestanding microtubes with clear insertable flat cap, no graduations, assembled pack0,5 mltrasparente+Log in for availability1 * 500 STSFr. 138.00Specifications:New ProductNew Product
Screw Cap Microtubes with Tethered Socket Caps in Separate Resealable Bags
Product Information
- DescrizioneCapacitàColoreSteriliAvailabilityPrice
- Conical base microtubes with clear tethered, socket cap, no graduations, combo pack0,5 mltrasparente-Log in for availability1 * 500 STSFr. 91.40Specifications:New ProductNew Product
- Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack0,5 mltrasparente-Log in for availability1 * 500 STSFr. 85.80Specifications:New ProductNew Product
- Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack1,5 mltrasparente-Log in for availability1 * 500 STSFr. 91.40Specifications:New ProductNew Product
- Conical base microtubes with clear tethered, socket cap, graduated, combo pack2,0 mltrasparente-Log in for availability1 * 500 STSFr. 91.40Specifications:New ProductNew Product
- Conical base microtubes with clear tethered, socket cap, graduated, combo pack1,5 mltrasparente-Log in for availability1 * 500 STSFr. 91.40Specifications:New ProductNew Product
- Freestanding microtubes with clear tethered, socket cap, graduated, combo pack2,0 mltrasparente-Log in for availability1 * 500 STSFr. 85.80Specifications:New ProductNew Product
Customisable Socket Caps
Product Information
- Catalog #DescrizioneAvailabilityPrice
Colour Inserts for Socket Caps
Product Information
- Catalog #DescrizioneColoreAvailabilityPrice