Orders LinkContactUsLinkComponent
MPC Coupling Caps
MPC Coupling Caps
New Product
Nome del fornitore:   Avantor Fluid Handling
MPC Coupling Caps
Nome del fornitore:  Avantor Fluid Handling
New Product
Certificates
MPC Coupling Caps
10 varianti
Product Details & Documents
MPC connectors are multi-purpose, open-format connectors for use in a wide variety of critical fluid handling applications. This particular type of connector is perfect for smaller bag systems for aliquoted media or other products stored in bags.
  • Ergonomic thumb latch: Easy to operate - even with gloved hands
  • USP Class VI materials: Meet biocompatibility
  • Sterilisable by autoclave, EtO, e-beam, or gamma: Robust and economical for single-use applications
  • Parting line-free hose barb: Eliminates potential leak path
  • BSE/TSE free materials: Meet ADCF requirements
MPC connectors add ease of use and security to the most critical fluid handling applications. Choose from a full line of male and female MPC connectors and configurations, including pressure sealing caps and plugs, in sizes to fit ¹/₈", ¹/₄", and ³/₈" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Specifications for Product Family
Specifiche
  • Pressione di esercizio massima:
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Up to 60 psi, 4,1 bar
  • Temperatura massima:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 149 °C (300 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 121 °C (250 °F)
  • Temperatura minima:
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • −40 °C (−40 °F)
  • Sterilizzazione:
  • Gamma, autoclave
  • Gamma, autoclave
  • Gamma, autoclave
  • Gamma, autoclave
  • Gamma, autoclave
  • Gamma, autoclave
  • Gamma, autoclave
  • Gamma, autoclave
Specifiche
Product Family Options
MPX
Informazioni sul prodotto
  • Tipo
    Dimensioni del raccordo/Ø int. del tubo (porta 1)
    Tipo di connessione
    Fitting material
    Flow pattern
    Disponibilità
    Price
  • MFLX30003-86
    ¹/₂" Hose barb non-valved coupling body PC, MPX17803B25; 25 Pk
    ¹/₂" Hose barb non-valved coupling body PC, MPX17803B25; 25 Pk
    Open
    ¹/₂"
    Quick-disconnect to barb
    PC
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - SFr. 113.00
    SFr. 113.00
    1 * 25 ST
       SFr. 113.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-86
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    121 °C (250 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
  • MFLX30003-87
    ³/₈" Hose barb non-valved coupling insert PC, MPX22603M; 250 Pk
    ³/₈" Hose barb non-valved coupling insert PC, MPX22603M; 250 Pk
    Open
    ³/₈"
    Quick-disconnect to barb
    PC
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 250 ST
    1 * 250 ST - SFr. 1’100.00
    SFr. 1’100.00
    1 * 250 ST
       SFr. 1’100.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-87
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    121 °C (250 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
  • MFLX30003-88
    MPX coupling plug PS, MPX30039MB25; 25 Pk
    MPX coupling plug PS, MPX30039MB25; 25 Pk
    Quick-disconnect plug
    PPSU
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - SFr. 329.00
    SFr. 329.00
    1 * 25 ST
       SFr. 329.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-88
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    149 °C (300 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
  • MFLX30003-89
    MPX coupling plug PC, MPX32003B25; 25 Pk
    MPX coupling plug PC, MPX32003B25; 25 Pk
    Quick-disconnect plug
    PC
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - SFr. 135.00
    SFr. 135.00
    1 * 25 ST
       SFr. 135.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-89
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    121 °C (250 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
MPXK
Informazioni sul prodotto
  • Tipo
    Dimensioni del raccordo/Ø int. del tubo (porta 1)
    Tipo di connessione
    Fitting material
    Flow pattern
    Disponibilità
    Price
  • MFLX30003-91
    MPX Coupling cap, locking PC, MPXK32003B25; 25 Pk
    MPX Coupling cap, locking PC, MPXK32003B25; 25 Pk
    Quick-disconnect cap
    PC
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - SFr. 206.00
    SFr. 206.00
    1 * 25 ST
       SFr. 206.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-91
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    121 °C (250 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
  • MFLX30003-92
    MPX Coupling cap, locking PS, MPXK32039; 250 Pk
    MPX Coupling cap, locking PS, MPXK32039; 250 Pk
    Quick-disconnect cap
    PPSU
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 250 ST
    1 * 250 ST - SFr. 5’380.00
    SFr. 5’380.00
    1 * 250 ST
       SFr. 5’380.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-92
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    149 °C (300 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
  • MFLX30003-93
    MPX Coupling cap, locking PS, MPXK32039B25; 25 Pk
    MPX Coupling cap, locking PS, MPXK32039B25; 25 Pk
    Quick-disconnect cap
    PPSU
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - SFr. 478.00
    SFr. 478.00
    1 * 25 ST
       SFr. 478.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-93
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    149 °C (300 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
  • MFLX30003-90
    ¹/₂" Hose barb non-valved coupling body with lock PC, MPXK17803B25; 25 Pk
    ¹/₂" Hose barb non-valved coupling body with lock PC, MPXK17803B25; 25 Pk
    Open
    ¹/₂"
    Quick-disconnect to barb
    PC
    Straight
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - SFr. 189.00
    SFr. 189.00
    1 * 25 ST
       SFr. 189.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-90
    Pressione di esercizio massima
    Up to 60 psi, 4,1 bar
    Temperatura massima
    121 °C (250 °F)
    Temperatura minima
    −40 °C (−40 °F)
    Sterilizzazione
    Gamma, autoclave
    New Product
Silicone Replacement Seals
Informazioni sul prodotto
  • Tipo
    Dimensioni del raccordo/Ø int. del tubo (porta 1)
    Tipo di connessione
    Fitting material
    Flow pattern
    Disponibilità
    Price
  • MFLX30003-00
    SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone
    SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone
    Silicone
    Log in per disponibilità
    Log in per disponibilità
    1 * 50 ST
    1 * 50 ST - SFr. 176.00
    SFr. 176.00
    1 * 50 ST
       SFr. 176.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-00
    Pressione di esercizio massima
    Temperatura massima
    Temperatura minima
    Sterilizzazione
    New Product
  • MFLX30003-01
    SaniQuik USP class VI, ADCF silicone seal, 1-¹/₂"; 50 Pk
    SaniQuik USP class VI, ADCF silicone seal, 1-¹/₂"; 50 Pk
    Silicone
    Log in per disponibilità
    Log in per disponibilità
    1 * 50 ST
    1 * 50 ST - SFr. 265.00
    SFr. 265.00
    1 * 50 ST
       SFr. 265.00
    Specifiche:
    New Product
    More
    Specifications
    Cat. No.
    MFLX30003-01
    Pressione di esercizio massima
    Temperatura massima
    Temperatura minima
    Sterilizzazione