Proteometer-CV MD buffer
Proteometer-CV MD Buffer Packets (4X)
Nome del fornitore: NOVILYTIC
Proteometer-CV MD buffer
Codice catalogo NOVIBUF-PCV-003
Nome del fornitore: NOVILYTIC
New Product
Numero CAS:
Vai a
Product Details & Documents
4 sealed buffer packets specifically formulated for use with Proteometer-CV kit.
Buffer packets available for reorder when the Proteometer-CV Reactor, MD Unit, and/or Proteometer-CV reagent are not required
Intended for use with the Proteometer-CV kit.
Informazioni per l'ordine: Reorder in sets of 4 as needed.
Confezione: Supplied in a custom box with instructions for use. Store at room temperature.
Specifiche
Specifiche
- Descrizione:Proteometer-CV MD Buffer Packets (4X)