Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

JK 184 ≥99% Hedgehog signaling inhibitor

Marketsource
Nome del fornitore:   Abcam
JK 184 ≥99% Hedgehog signaling inhibitor
Nome del fornitore:  Abcam
Marketsource
Certificates

2 varianti

Product Details & Documents

Potent Hedgehog (Hh) signaling inhibitor. Inhibits alcohol dehydrogenase 7 (Adh7) (IC₅₀ = 210 nM) and prevents Gli-dependent transcriptional activity (IC₅₀ = 30 nM). Potently inhibits microtubule assembly. Additionally displays antitumor activity.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Sinonimi:  JK184, N-(4-Ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)thiazol-2-amine
  • Numero CAS:315703-52-7
  • Molecular Weight:350,44 g/mol
Specifications for Product Family

Risultati dei test sulle specifiche

  • Constituents::
    100% JK184
Product Family Options

Informazioni sul prodotto

  • Disponibilità
    Price
  • Log in per disponibilità
    Log in per disponibilità
    1 * 10 mg
    1 * 10 mg - SFr. 180.00
    SFr. 180.00
    1 * 10 mg
       SFr. 180.00

    Specifiche:
    Marketsource
    Marketsource
  • Log in per disponibilità
    Log in per disponibilità
    1 * 50 mg
    1 * 50 mg - SFr. 764.00
    SFr. 764.00
    1 * 50 mg
       SFr. 764.00

    Specifiche:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.