Thymol ≥98% phenolic disinfectant
Product Details & Documents
Phenolic compound, originally obtained from thyme oil, that is used to inhibit oral bacteria. Also used as a cosmetic biocide/preservative and/or fragrance ingredient. Functions as a denaturant and fragrance ingredient in cosmetic formulations.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Numero CAS:89-83-8
- Molecular Weight:150,22 g/mol
Specifiche
Risultati dei test sulle specifiche
- Constituents::100% Thymol
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



