Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

D-Cycloserine ≥99% NMDA glycine site agonist

Nome del fornitore:   Abcam
D-Cycloserine ≥99% NMDA glycine site agonist
Nome del fornitore:  Abcam
Certificates

2 varianti

Product Details & Documents

Partial agonist at the NMDA receptor glycine recognition site. Enhances learning and memory in vivo. Perfomance enhancer in a variety of cognitive models. Also available in simple stock solutions - add 1 ml of water to get an exact, ready-to-use concentration.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Sinonimi:  D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
  • Numero CAS:68-41-7
  • Molecular Weight:102,09 g/mol
Specifications for Product Family

Risultati dei test sulle specifiche

  • Constituents::
    100% (R)-4-amino-1,2-oxazolidin-3-one
Product Family Options

Informazioni sul prodotto

  • Disponibilità
    Price
  • Log in per disponibilità
    Log in per disponibilità
    1 * 1 g
    1 * 1 g - SFr. 89.80
    SFr. 89.80
    1 * 1 g
       SFr. 89.80

  • Log in per disponibilità
    Log in per disponibilità
    1 * 5 g
    1 * 5 g - SFr. 300.00
    SFr. 300.00
    1 * 5 g
       SFr. 300.00