Orders LinkContactUsLinkComponent
Actinonin ≥97% Matrix metalloprotease (MMP) inhibitor
 :   Abcam
Actinonin ≥97% Matrix metalloprotease (MMP) inhibitor
 :  Abcam
Actinonin ≥97% Matrix metalloprotease (MMP) inhibitor
Potent MMP and peptide deformylase inhibitor. Shows potent activity against peptide deformylase, M, N, leucine, alanyl, dipeptidyl aminopeptidases, enkephalinase A, and meprin α and β. Shows antiproliferative effects. Shows male-specific anti-ischemic effects in vivo.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
  •  :13434-13-4
  •  :385,5 g/mol
  • Constituents::
    100% Actinonin
Product Family Options
  •  
  •    
    1 * 5 mg
    1 * 5 mg - SFr. 105.00
    SFr. 105.00
    1 * 5 mg
       SFr. 105.00
  •    
    1 * 25 mg
    1 * 25 mg - SFr. 424.00
    SFr. 424.00
    1 * 25 mg
       SFr. 424.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.