Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Reserpine ≥98% Irreversible vesicular monoamine transporter antagonist

Reserpine ≥98% Irreversible vesicular monoamine transporter antagonist

Marketsource
Anbieter:   Abcam
Reserpine ≥98% Irreversible vesicular monoamine transporter antagonist
Anbieter:  Abcam
Marketsource
Zertifikate
Reserpine ≥98% Irreversible vesicular monoamine transporter antagonist

2 Varianten

Product Details & Documents

Irreversible vesicular monoamine transporter (VMAT2) antagonist. Long-lasting bioamine depleter. Prevents bioamine transport into synaptic vesicles and chromaffin granules. Depletes biogenic amine stores. Antihypertensive and antipsychotic.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Nummer:50-55-5
  • Molekulargewicht:608,68 g/mol
Spezifikationen für diese Produktfamilie

Ergebnisse des Spezifikationstests

  • Constituents::
    100% Reserpine
Product Family Options

Produktinformationen

  • Verfügbarkeit
    Preis
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 250 mg
    1 * 250 mg - SFr. 69.90
    SFr. 69.90
    1 * 250 mg
       SFr. 69.90

    Spezifikationen:
    Marketsource
    Marketsource
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 g
    1 * 1 g - SFr. 145.00
    SFr. 145.00
    1 * 1 g
       SFr. 145.00

    Spezifikationen:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.