Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Ampicillin trihydrate ≥98% beta-lactam antibiotic

Supplier:   Abcam
Ampicillin trihydrate ≥98% beta-lactam antibiotic
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

β-lactam antibiotic (MIC₅₀ =3,12 μg/ml). Competitively inhibits transpeptidase causing cell wall disruption to Gram-positive and Gram-negative bacteria. Active in vitroand in vivo.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
  • CAS Number:69-53-4
  • Molecular Weight:349,4 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Ampicillin trihydrate
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 79.80
    SFr. 79.80
    1 * 5 g
       SFr. 79.80

unology General

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 275.00
    SFr. 275.00
    1 * 25 g
       SFr. 275.00