Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
ChIP Kit, Abcam

ChIP Kit, Abcam

ChIP Kit
Codice catalogo ABCAAB500-1
Marketsource
Nome del fornitore:   Abcam
ChIP Kit, Abcam
Codice catalogo ABCAAB500-1
Nome del fornitore:  Abcam
Marketsource
Numero CAS:  
ChIP Kit, Abcam

ChIP Kit

Numero CAS:  
Price
SFr. 609.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents

Chromatin immunoprecipitation (ChIP) kit. Suitable for mammalian suspension and adherent cells. - Cited in over 165 publications - Includes reagents for chromatin shearing & extraction - qPCR-ready DNA purification - Uses protein A sepharose for antibody pulldown

Attenzione: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Descrizione:
    ChIP Kit
  • Cat. No.:
    ABCAAB500-1
  • Temperatura di conservazione:
    Store at Multi.

Specifiche