Orders LinkContactUsLinkComponent
RNase R
RNase R
Codice catalogo ABCAAB286929-1KIT
Nome del fornitore:   Abcam
RNase R
Codice catalogo ABCAAB286929-1KIT
Nome del fornitore:  Abcam
Numero CAS:  
Numero CAS:  
Price
SFr. 360.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
RNase R is an E. coli exoribonuclease that exhibits 3'-to-5' exonuclease activity, efficiently digesting nearly all linear RNA species. - Efficiently digests linear RNA species with 3'-to-5' exonuclease activity - High purity – contains RNase R at 10 U/µl for optimal performance
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifiche
Specifiche
  • Cat. No.:
    ABCAAB286929-1KIT
  • Nome dell'enzima:
    RNase R
Specifiche