Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
ALLN

ALLN

Marketsource
Nome del fornitore:   Abcam
ALLN
Nome del fornitore:  Abcam
Marketsource
Certificates

2 varianti

Product Details & Documents

MW 383.5 Da, Purity >95%. ALLN, cysteine proteinase inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.

  • Constituents: 100% ALLN

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Specifiche

Specifiche

  • Catalog No:
  • ABCAAB141445-5
  • ABCAAB141445-25
  • Protein/peptide name:
  • ALLN
  • ALLN
  • CAS No.:
  • 5 mg
  • Purezza:
  • >95%
  • >95%
  • Peso molecolare:
  • 383.5 DA
  • 383.5 DA
  • Condizioni di conservazione:
  • Store at -20 °C.
  • Store at -20 °C.
  • Temperatura di spedizione:
  • Ambient - Can Ship with Ice
  • Ambient - Can Ship with Ice

Specifiche

Product Family Options

Informazioni sul prodotto

  • Disponibilità
    Price
  • Log in per disponibilità
    Log in per disponibilità
    1 * 5 mg
    1 * 5 mg - SFr. 180.00
    SFr. 180.00
    1 * 5 mg
       SFr. 180.00

    Specifiche:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB141445-5
    Protein/peptide name
    ALLN
    CAS No.
    5 mg
    Purezza
    >95%
    Peso molecolare
    383.5 DA
    Condizioni di conservazione
    Store at -20 °C.
    Temperatura di spedizione
    Ambient - Can Ship with Ice
    Marketsource

unology General

Informazioni sul prodotto

  • Disponibilità
    Price
  • Log in per disponibilità
    Log in per disponibilità
    1 * 25 mg
    1 * 25 mg - SFr. 694.00
    SFr. 694.00
    1 * 25 mg
       SFr. 694.00

    Specifiche:
    Marketsource
    More
    Specifications
    Cat. No.
    ABCAAB141445-25
    Protein/peptide name
    ALLN
    CAS No.
    Purezza
    >95%
    Peso molecolare
    383.5 DA
    Condizioni di conservazione
    Store at -20 °C.
    Temperatura di spedizione
    Ambient - Can Ship with Ice
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.