Orders LinkContactUsLinkComponent
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
Marketsource
Nome del fornitore:   Revvity
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
Nome del fornitore:  Revvity
Marketsource
Certificates
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
3 varianti
Product Details & Documents
The NEXTFLEX® Rapid XP V2 DNA-sequence kit offers a simplified DNA-seq library prep workflow for Illumina® platforms. Libraries produced using this kit are normalised during library construction.
  • Fully automated normalization and fragmentation Enzymatic fragmentation with end-repair and A-tailing in one reaction Highly efficient ligation with low adapter dimer formation Efficient: Automated on the Sciclone® and Zephyr® NGS workstations for mid-high throughput automation and now on the BioQule™ NGS System for low throughput labs
The NEXTFLEX® Rapid XP V2 DNA-seq kit includes proprietary NEXTFLEX® normalization beads, which provide a consistent mass and sequencing cluster density for all samples in a library pool. This shortens the time needed for quantification and pooling preparation for sequencing in a high throughput lab by up to 3 hours per 96 samples. The NEXTFLEX® Rapid XP V2 DNA-seq kit offers a streamlined workflow with highly efficient ligation, generating low amount of adapter dimers even at low input concentrations.
The NEXTFLEX Rapid XP V2 DNA-Seq kit is compatible with multiple liquid handlers, including the Sciclone G3 NGSx workstation, Sciclone G3 NGSx iQ™ workstation, and Zephyr NGS workstation. For low throughput automation, the NEXTFLEX Rapid XP V2 DNA-Seq kit is also now automated on the BioQule™ NGS System which simplifies both library prep and quantitation. This low-cost, benchtop instrument delivers libraries ready to load into your sequencer with only 15 minutes of hands-on time.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Informazioni sul prodotto
  • No. of reactions
    Disponibilità
    Price
  • BIOONOVA-5149-21
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    8
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 KIT
    1 * 1 KIT - SFr. 303.00
    SFr. 303.00
    1 * 1 KIT
       SFr. 303.00
    Specifiche:
    Marketsource
    Marketsource
  • BIOONOVA-5149-22
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    48
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 KIT
    1 * 1 KIT - SFr. 1’806.00
    SFr. 1’806.00
    1 * 1 KIT
       SFr. 1’806.00
    Specifiche:
    Marketsource
    Marketsource
  • BIOONOVA-5149-23
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
    96
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 KIT
    1 * 1 KIT - SFr. 2’193.00
    SFr. 2’193.00
    1 * 1 KIT
       SFr. 2’193.00
    Specifiche:
    Marketsource