Tork T4 Extra Soft Conventional Toilet Paper Roll, White
Codice catalogo SCAA110319
Nome del fornitore: Essity
New Product
Specifiche
- Confezione:7 packs, 8 rolls each. 155 sheets each, 3 ply.
- Descrizione:Premium extra soft conventional toilet paper roll, refill, white, compatible with Tork T4 dispensers
- L×L:19100×98 mm
Specifiche
Informazioni su questo articolo
Tork T4 extra soft conventional toilet paper roll, provides a luxurious look and feel with superior performance for upscale washrooms in hotels or office buildings.
Avoid waste, thanks to the perforated sheets that enable efficient tear-off and use
-
Save time on refills with Tork Easy Handling® packaging, it makes carrying and disposing of packaging effortless
Universal quality
Sheet length 123 mm
Core inner diameter 38 mm