Tork Xpress® H2 Compressed Soft Multifold Hand Towels, White
Codice catalogo SCAA100888
Nome del fornitore: Essity
New Product
Specifiche
- Per:Compatible with H2 dispensers
- Confezione:12 packs, 170 paper hand towels each; 2 ply
- Colore:bianco
- Descrizione:Premium multifold hand towels, refill
- L×L:320×212 mm (Unfolded size); 8 mm L (Folded size)
Specifiche
Informazioni su questo articolo
Tork Xpress® compressed soft multifold hand towels white H2, premium, 2-ply, M-fold, large. Bring extra comfort to guests in high-end establishments with these soft, thick and large hand towels.
Reduce paper towel consumption in washrooms and improve hygiene with one-at-a-time dispensing
- Reduce waste and increase cost savings - these hand towels are quick-drying and absorbent, so guests don't use as many
- Maximise the space - use these compressed paper hand towels with the compact Tork Xpress® dispenser in low-to-medium traffic washrooms
-
Premium quality
Compatible with H2 dispensers
Save time restocking dispensers with 50% compressed paper hand towels that double dispenser capacity. Guests enjoy a premium experience with these soft and absorbent hand towels. One-at-a-time dispensing helps improve hygiene and reduce waste as guests only touch the towel they take. Our compact dispensers fit into small spaces for maximum convenience.
Certifications:
EU Ecolabel, SE/004/001; FSC MIX Credit, SA-COC-008266.