Tork PeakServe® H5 Mini Continuous™ Paper Hand Towel Dispenser, Black
Catalog # SCAA552558
Supplier: Essity
New Product
Specifications
- Colore:nero
- Descrizione:Continuous hand towel dispenser
- L×P×A:101×367×491 mm
Specifications
About this item
Tork PeakServe® Mini Continuous™ paper hand towel dispenser black H5, high capacity, elevation range.
Prevent washroom queues for paper towels - the Tork PeakServe® paper towel dispenser serves guests in just three seconds.
- Never run out of paper towels and cut refilling time in half thanks to the dispenser's high capacity of 1230 towels
- Save space with paper towel bundles compressed by 50% fitting more in storerooms, on cleaning trolleys and in the dispenser
- Don't waste time on maintenance - thanks to a flexible refill system, users can refill Tork paper towels when it suits them
-
Material: Plastic
Suitable for H5 refills from Tork
High-traffic washrooms can be challenging to maintain, with paper hand towel shortages, jams and the need for frequent refilling creating long waits for guests. It cuts refill time in half and eliminates paper jams thanks to the patented Continuous™ hand towel system - and all this in an extra slim ADA-compliant dispenser that is perfect for compact spaces. Carbon neutral - produced with certified renewable electricity and compensated with climate projects.