- Colore:nero
- Descrizione:Continuous hand towel dispenser
- L×P×A:101×370×731 mm
Tork PeakServe® Continuous™ paper hand towel dispenser black H5, high capacity, elevation range.
Say goodbye to queues in busy washrooms with this paper towel dispenser that delivers a towel in just three seconds
- Prevent hand towels running out with 250% more capacity than Tork Xpress multifold paper towel dispensers
- Cut refilling time in half and spend more time on other tasks with a hand towel dispenser, can be refilled at any point
-
Avoid towel jams or towel tabbing with the patented Continuous™ system for paper towels
Material: Plastic
Suitable for H5 refills from Tork
Avoid washroom traffic jams and paper towel run-outs with this paper hand towel dispenser. Perfect for high-traffic washrooms, it uses compressed hand towels to deliver 250% more capacity in a slim, ADA-compliant design. Save staff time with the flexible refill system that allows them to refill the dispenser at any time.
Improve the flow of people in busy washroom with the patented Continuous™ system that smoothly dispenses Tork paper towels in just three seconds. This system is certified easy-to-use by the Swedish Rheumatism Association (SRA) making hygiene accessible for all. Carbon neutral - produced with certified renewable electricity and compensated with climate projects.