Tork S5 Mildly Scented Hand Washing Mini Foam Soap
Codice catalogo SCAA525502
Nome del fornitore: Essity
New Product
Specifiche
- Confezione:8 cans per box, 525 ml each
- Descrizione:Premium foam soap, refill
Specifiche
Informazioni su questo articolo
Tork mildly scented hand washing mini foam soap S5, contains moisturising ingredients. It is dermatologically tested and fresh-scented foam hand soap.
A gentle soap that helps prevent dry skin with moisturising, lipid-replenishing ingredients and a pearly shine
- 96% of ingredients are from natural origin (according to specification and ISO 16128, including water)
- Minimise energy use - this soap is proven effective in cold water (based on testing at 20 °C)
-
Fresh scent fragrance, premium quality
Colour: Transparent
Compatible with S5 dispensers
Provide care for normal and dry skin for everyday hand wash. Tork mildly scented foam soap has a fresh scent, plus moisturising and replenishing ingredients, and a soft and creamy lather for gentle hand washing.
Suitable for Tork mini soap and sanitiser dispensers, this soap is certified easy-to-use and provides good hand hygiene to all users.
Certifications:
EU Ecolabel, SE/030/002.