Tork S5 Mildly Scented Hand Washing Mini Liquid Soap
Katalognr. SCAA425502
Anbieter: Essity
New Product
Technische Daten
- Confezione:8 cans per box, 525 ml each
- Descrizione:Premium liquid soap, refill
Technische Daten
Über diesen Artikel
Tork mildly scented hand washing mini liquid soap S5, contains moisturising ingredients, dermatologically tested.
A gentle soap with moisturising, lipid-replenishing ingredients and a pearly shine
- 96% of ingredients are from natural origin (according to specification and ISO 16128, including water)
- Minimise energy use - this soap is proven effective in cold water (based on testing at 20 °C)
-
Fresh Scent fragrance, premium quality
Colour: Light yellow
Compatible with S5 dispensers
Provide care for normal and dry skin for everyday hand wash. Tork mildly scented liquid soap has a fresh scent, plus moisturising and replenishing ingredients for gentle hand washing. Suitable for Tork mini soap and sanitiser dispensers, this soap is certified easy-to-use and provides good hand hygiene to all users.
Certifications:
EU Ecolabel, SE/030/002.