Tork S5 Luxury Hand Washing Mini Foam Soap
Catalog # SCAA525902
Supplier: Essity
New Product
Specifications
- Confezione:8 cans per box, 525 ml each
- Descrizione:Premium foam soap, refill
Specifications
About this item
Enriched with natural magnolia extract for a rich, creamy hand-washing experience.
Minimise energy use - this soap is proven effective in cold water (based on testing at 20 °C)
- Fast refilling in less than 10 seconds
- Helps reduce water consumption by over 10% (comparing 2000 handwashes with one dose of Tork luxury foam soap vs Tork mildly scented liquid soap)
-
Floral fragrance, premium quality
Color: Transparent
Compatible with S5 dispensers
Indulge visitors with an extra touch of luxury with Tork luxury foam soap. A luxuriously scented formulation enriched with natural magnolia extract and moisturising, lipid-replenishing ingredients for an indulgent, creamy hand-washing experience.
Suitable for Tork mini soap and sanitiser dispensers, this foam soap is proven easy to use and provides good hand hygiene for all users.
Certifications:
EU Ecolabel, SE/030/002.