Tork S4 Odour-Control Liquid Soap
Catalog # SCAA424011
Supplier: Essity
New Product
Specifications
- Confezione:6 cans per box, 1000 ml each
- Descrizione:Premium liquid soap, refill
Specifications
About this item
Helps remove common food odours from the hands, making it ideal for use in a professional kitchen.
No fragrance or colour added means this dermatologically tested formula with a skin-friendly pH is gentle to the skin
- Save staff time with effortless refilling - certified by the Swedish Rheumatism Association, tested according to the Design for All test, published in the Swedish Design Research Journal, 2011
- Help secure good hand hygiene and reduce the risk of contamination as the factory-sealed bottle has a new pump for every refill and is collapsible for 70% less waste volume (based on Essity test)
-
Perfume free fragrance, premium quality
Color: Transparent
Compatible with S4 dispensers
Perfect for professional kitchens, Tork odour-control liquid soap removes common food odours from hands. It is formulated without added fragrance or colour and is dermatologically tested. This foam soap is suitable for Tork S4 dispensers, is certified easy-to-use and provides good hand hygiene thanks to a factory-sealed refill bottle.
Certifications:
EU Ecolabel, SE/030/002.