ContactUsLinkComponent
Tork Reflex™ M3 Wiping Paper Towels
Tork Reflex™ M3 Wiping Paper Towels
Catalog # SCAA473246
Supplier:  Essity
New Product
undefined
Tork Reflex™ M3 Wiping Paper Towels
Catalog # SCAA473246
Supplier:  Essity
New Product
Specifications
  • Confezione:
    12×343 sheets
  • Colore:
    bianco
  • Descrizione:
    Centerfeed reflex, refill, advanced, compatible with Tork M3 dispensers
Specifications
About this item
Tork OptiServe® Coreless is a high-capacity coreless toilet paper system for facilities where efficiency, sustainability and providing a great experience are key. High capacity, reduced storage space, and a certified easy to use design make it a better alternative to conventional toilet paper and jumbo rolls.
  • Cut down consumption by up to 37% with single-sheet dispensing, which makes it easy to complete one-handed tasks
    Improve hygiene with a food contact approved roll - only what is taken is touched
    No lengthy maintenance, thanks to SmartCore® core removal - fast and easy refilling frees up time
    Avoid using multiple rolls with the multipurpose wiping paper that's strong and absorbent
Wiping paper can break down easily and generate waste. But strong Reflex™ wiping paper uses single-sheet dispensing to reduce consumption by up to 37% compared with freeflow Tork wiping paper Plus. Improve hygiene levels with the food contact approved industrial paper roll  - sheets can be easily taken one-handed from a centrefeed dispenser, so only what is taken is touched. The SmartCore® core removal and run out indicator work to avoid time-consuming refills and empty dispensers.
Certifications: FSC Recycled credit, SA-COC-008266; EU Ecolabel, SE/004/001.