ContactUsLinkComponent
Sartobind® Lab LC System Adapter Kit
Sartobind® Lab LC System Adapter Kit
Catalog # SARTSBLAAU01-1
Supplier:  Sartorius
New Product
undefined
Sartobind® Lab LC System Adapter Kit
Catalog # SARTSBLAAU01-1
Supplier:  Sartorius
New Product
Specifications
  • Descrizione:
    Sartobind® Lab LC system adapter kit, 60×80×10 mm
Specifications
About this item
Sartobind® Rapid A Lab incorporates the state-of-the-art in membrane affinity chromatography (AC) technology, offering fast, flexible and future-proof antibody purification. It is an ideal solution for antibody research, as well as downstream process development for novel biologics before starting clinical and commercial production.
  • The fastest matrix. High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes
  • Productivity boost. Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity
  • Plug and play. Collapse your workflow. Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration
  • Flexibility built in. Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system
  • Process-ready technology. Seamless scalability. Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production