Orders LinkContactUsLinkComponent
Sartobind® Rapid A Labs
Sartobind® Rapid A Labs
   Opzioni sostenibili disponibili
Nome del fornitore:   Sartorius
Sartobind® Rapid A Labs
Nome del fornitore:  Sartorius
   Opzioni sostenibili disponibili
Certificates
Sartobind® Rapid A Labs
2 varianti
Product Details & Documents
To affinity and beyond: Fast, flexible and future-proof antibody purification.
  • The fastest matrix. High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes
  • Productivity boost. Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity
  • Plug and play. Collapse your workflow. Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration
  • Flexibility built in. Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system
  • Process-ready technology. Seamless scalability. Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production
Sartobind® Rapid A Lab incorporates the state-of-the-art in membrane affinity chromatography (AC) technology, offering fast, flexible and future-proof antibody purification. It is an ideal solution for antibody research, as well as downstream process development for novel biologics before starting clinical and commercial production.
Bed volume: 0,5 ml
Ligand: Protein A
Membrane support: Polyethylene terephthalate (PET)
Specifications for Product Family
Specifiche
  • Catalog No:
  • SARTSBLRA025EL-1
  • SARTSBLRA025EL-A
  • Environmentally Preferable:
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Certificazione ambientale:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifiche
Product Family Options
Informazioni sul prodotto
  • Disponibilità
    Price
  • SARTSBLRA025EL-1
    Sartobind® Rapid A lab, 125×120×62 mm
    Sartobind® Rapid A lab, 125×120×62 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 211.00
    SFr. 211.00
    1 * 1 ST
       SFr. 211.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    SARTSBLRA025EL-1
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate
      Produzione a basso impatto
  • SARTSBLRA025EL-A
    Sartobind® Rapid A lab, 203×158×78 mm
    Sartobind® Rapid A lab, 203×158×78 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 4 ST
    1 * 4 ST - SFr. 727.00
    SFr. 727.00
    1 * 4 ST
       SFr. 727.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    SARTSBLRA025EL-A
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate
      Produzione a basso impatto
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Sartobind® Lab LC system adapter kit, 60×80×10 mm
    Sartobind® Lab LC system adapter kit, 60×80×10 mm
    SARTSBLAAU01-1
    Log in per disponibilità
    1 * 1 ST - SFr. 31.00