PYREX® Roller Bottle
Roller bottle, GL 45 thread, Ø ext. 110 mm, growth area: 670 cm² (effective), 820 cm² (overall)
Codice catalogo BIBB1528/02
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
Designed for use in conjunction with roller apparatus, these PYREX® roller bottles are ideal for cultivating large quantities of anchorage dependent cells.
Ideal for cell culture work
- Wide neck for easy cell harvesting
- Non-toxic polypropylene screw cap
- Autoclavable up to 121 °C
- Fits standard roller apparatus
PYREX® borosilicate glass construction is specially selected for surface finish and optical clarity. This bottle can be used with membrane screw caps to allow gaseous transfer.
Specifiche
- Descrizione:Roller bottle, GL 45 thread, Ø ext. 110 mm, growth area: 670 cm² (effective), 820 cm² (overall)
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..