ContactUsLinkComponent
Milli-Q SQPAK™ TOC Quanta Cartridge
Milli-Q SQPAK™ TOC Quanta Cartridge
Catalog # MILLSQPAKT0CM1
Supplier:  Merck
New Product
undefined
Milli-Q SQPAK™ TOC Quanta Cartridge
Catalog # MILLSQPAKT0CM1
Supplier:  Merck
New Product
Specifications
  • Descrizione:
    SQPAK™ TOC Quanta Cartridge, produces ultrapure (Type 1) water with TOC ≤ 5 ppb for use with the ultrapure dispensing module of Milli-Q® SQ 2Series systems
Specifications
About this item
The SQPAK™ TOC Quanta cartridge produces ultrapure (Type 1) water at 18,2 MΩ·cm resistivity and TOC ≤5 ppb. The cartridge contains IQnano™ ion-exchange media to remove ions down to trace levels and high-grade synthetic activated carbon that targets traces of organic contaminants.
  • Easy replacement thanks to the Twist and Lock mechanism
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
Ultrapure label and blue colour of the SQPAK™ Quanta cartridge are repeated on the production station interface, to inform about its water purification purpose and guide replacement.