ContactUsLinkComponent
Milli-Q SQPAK™ Quanta Cartridge
  MILLSQPAKQUAM1
 :  Merck
New Product
Milli-Q SQPAK™ Quanta Cartridge
  MILLSQPAKQUAM1
 :  Merck
New Product
  • Descrizione:
    SQPAK™ Quanta Cartridge, produces ultrapure (Type 1) water for use on the ultrapure dispensing module of Milli-Q® SQ 2Series systems
SQPAK™ Quanta cartridge is a high-capacity polishing cartridge that produces ultrapure (Type 1) water at 18,2 MΩ·cm resistivity. The cartridge contains IQnano™ ion-exchange media to remove ions down to trace levels.
  • Easy replacement thanks to the Twist and Lock mechanism
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
Ultrapure label and blue colour of the SQPAK™ Quanta cartridge are repeated on the production station interface, to inform about its water purification purpose and guide replacement.