Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant TRP14 (from E. coli)

Human Recombinant TRP14 (from E. coli)

Codice catalogo ABCAAB236190-100
Marketsource
Nome del fornitore:   Abcam
Human Recombinant TRP14 (from E. coli)
Codice catalogo ABCAAB236190-100
Nome del fornitore:  Abcam
Marketsource
Numero CAS:  
Human Recombinant TRP14 (from E. coli)
Human Recombinant TRP14 (from E. coli)
Numero CAS:  
Price
SFr. 574.00
Log in per disponibilità
1 * 100 µG
Product Details & Documents

Recombinant Human TRP14 protein (Tagged) is a Human Full Length protein, in the 1 to 123 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Senza animali:
    N
  • Protein/peptide name:
    TRP14
  • Purezza:
    >90%
  • Attività biologica:
    Standard
  • Protein synonyms:
    TXNL5|TXNDC17|Thioredoxin domain-containing protein 17|14 kDa thioredoxin-related protein|Protein 42-9-9|Thioredoxin-like protein 5
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Sorgente luminosa:
    E. coli
  • Cat. No.:
    ABCAAB236190-100
  • Condizioni di conservazione:
    Store at −20 °C
  • Protein/peptide type:
    Recombinant
  • Forma:
    Liquid
  • Applicazioni testate:
    SDS-PAGE
  • Formulazione:
    pH: 7,2 - 7,4; Constituents: Tris buffer, 50% Glycerol (glycerin, glycerine)
  • Temperatura di spedizione:
    20

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.