Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant M6PR (cation dependent) (from E. coli)

Human Recombinant M6PR (cation dependent) (from E. coli)

Codice catalogo ABCAAB180308-50
Marketsource
Nome del fornitore:   Abcam
Human Recombinant M6PR (cation dependent) (from E. coli)
Codice catalogo ABCAAB180308-50
Nome del fornitore:  Abcam
Marketsource
Numero CAS:  
Human Recombinant M6PR (cation dependent) (from E. coli)
Human Recombinant M6PR (cation dependent) (from E. coli)
Numero CAS:  
Price
SFr. 734.00
Log in per disponibilità
1 * 50 µG
Product Details & Documents

Recombinant Human M6PR (cation dependent) protein - BSA and Azide free is a Human Fragment protein, in the 27 to 185 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Senza animali:
    N
  • Protein/peptide name:
    M6PR (cation dependent)
  • Purezza:
    >85%
  • Attività biologica:
    Standard
  • Protein synonyms:
    MPR46|MPRD|M6PR|Cation-dependent mannose-6-phosphate receptor|CD Man-6-P receptor|CD-MPR|46 kDa mannose 6-phosphate receptor|MPR 46
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Sorgente luminosa:
    E. coli
  • Cat. No.:
    ABCAAB180308-50
  • Condizioni di conservazione:
    Store at −20 °C
  • Protein/peptide type:
    Recombinant
  • Forma:
    Liquid
  • Applicazioni testate:
    SDS-PAGE
  • Formulazione:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,88% Sodium chloride, 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Temperatura di spedizione:
    20
  • Concentrazione:
    0,5 mg/ml

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..