Order Entry
Switzerland
ContactUsLinkComponent
 
Rat Recombinant ErbB2/HER2 (from E. coli)
  ABCAAB191967-10
 :  Abcam
 :  
undefined
Rat Recombinant ErbB2/HER2 (from E. coli)
  ABCAAB191967-10
 :  Abcam
 :  

 

  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    E. coli
  • Specie:
    Rat
  • Seq:
    1106
  • Condizioni di conservazione:
    Store at –20 °C
  • Contenuto di endotossine:
    <1 EU/µg
  • Attività biologica:
    Standard
  • Forma:
    Lyophilised
  • Senza animali:
    N
  • Protein synonyms:
    CD340|Neu|Erbb2|Receptor tyrosine-protein kinase erbB-2|Epidermal growth factor receptor-related protein|Proto-oncogene Neu|Proto-oncogene c-ErbB-2|p185erbB2|p185neu
  • Protein/peptide name:
    ErbB2/HER2
  • Purezza:
    >95%
  • Formulazione:
    pH: 7,4 Constituents: 24% Urea, 0,87% Sodium chloride, 0,24% Tris
  • Temperatura di spedizione:
    20
  • Applicazioni testate:
    HPLC, SDS-PAGE

 

 

Recombinant Rat ErbB2/HER2 protein is a Rat Fragment protein, in the 67 to 323 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.