Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant Corin (from Wheat germ)

Human Recombinant Corin (from Wheat germ)

Codice catalogo ABCAAB153116-10
Marketsource
Nome del fornitore:   Abcam
Human Recombinant Corin (from Wheat germ)
Codice catalogo ABCAAB153116-10
Nome del fornitore:  Abcam
Marketsource
Numero CAS:  
Human Recombinant Corin (from Wheat germ)
Human Recombinant Corin (from Wheat germ)
Numero CAS:  
Price
SFr. 634.00
Log in per disponibilità
1 * 10 µG
Product Details & Documents

Recombinant Human Corin protein is a Human Fragment protein, in the 616 to 715 aa range, expressed in Wheat germ and suitable for ELISA, WB.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Senza animali:
    N
  • Protein/peptide name:
    Corin
  • Attività biologica:
    Standard
  • Protein synonyms:
    CRN|TMPRSS10|CORIN|Atrial natriuretic peptide-converting enzyme|Corin|Heart-specific serine proteinase ATC2|Pro-ANP-converting enzyme|Transmembrane protease serine 10
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Sorgente luminosa:
    Wheat germ
  • Cat. No.:
    ABCAAB153116-10
  • Condizioni di conservazione:
    Store at −80 °C
  • Protein/peptide type:
    Recombinant
  • Forma:
    Liquid
  • Applicazioni testate:
    ELISA, WB
  • Formulazione:
    pH: 8, Constituents: 0,79% Tris HCl, 0,31% Glutathione
  • Temperatura di spedizione:
    Dry Ice

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..