Human Recombinant MARCH2 (from E. coli)
Informazioni su questo articolo
Recombinant human MARCH2 protein (His tag) is a Human Full Length protein, in the 1 to 246 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
2 varianti
Product Details & Documents
Recombinant human MARCH2 protein (His tag) is a Human Full Length protein, in the 1 to 246 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- ABCAAB314524-20UG
- ABCAAB314524-100UG
- Protein/peptide name:
- MARCH2
- MARCH2
- Protein synonyms:
- MARCH2|RNF172|HSPC240|MARCHF2|E3 ubiquitin-protein ligase MARCHF2|Membrane-associated RING finger protein 2|Membrane-associated RING-CH protein II|RING finger protein 172|RING-type E3 ubiquitin transferase MARCHF2|MARCH-II
- MARCH2|RNF172|HSPC240|MARCHF2|E3 ubiquitin-protein ligase MARCHF2|Membrane-associated RING finger protein 2|Membrane-associated RING-CH protein II|RING finger protein 172|RING-type E3 ubiquitin transferase MARCHF2|MARCH-II
- Protein/peptide type:
- Recombinant
- Recombinant
- Specie:
- Human
- Human
- Sorgente luminosa:
- E. coli
- E. coli
- Coniugazione:
- Non coniugato
- Non coniugato
- Senza animali:
- Y
- Y
- Attività biologica:
- Standard
- Standard
- Purezza:
- 90%
- 90%
- Forma:
- Lyophilised
- Lyophilised
- Formulazione:
- pH: 7,4 - 8, Constituents: 6% Trehalose, 0,14% PBS
- pH: 7,4 - 8, Constituents: 6% Trehalose, 0,14% PBS
- Condizioni di conservazione:
- Store at −20 °C
- Store at −20 °C
- Temperatura di spedizione:
- 20
- 20
- Applicazioni testate:
- SDS-PAGE
- SDS-PAGE
Specifiche
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- Specifiche:New Product
More
Specifications
Cat. No.ABCAAB314524-20UGProtein/peptide nameMARCH2Protein synonymsMARCH2|RNF172|HSPC240|MARCHF2|E3 ubiquitin-protein ligase MARCHF2|Membrane-associated RING finger protein 2|Membrane-associated RING-CH protein II|RING finger protein 172|RING-type E3 ubiquitin transferase MARCHF2|MARCH-IIProtein/peptide typeRecombinantSpecieHumanSorgente luminosaE. coliConiugazioneNon coniugatoSenza animaliYAttività biologicaStandardPurezza90%FormaLyophilisedFormulazionepH: 7,4 - 8, Constituents: 6% Trehalose, 0,14% PBSCondizioni di conservazioneStore at −20 °CTemperatura di spedizione20Applicazioni testateSDS-PAGENew Product - Specifiche:New Product
More
Specifications
Cat. No.ABCAAB314524-100UGProtein/peptide nameMARCH2Protein synonymsMARCH2|RNF172|HSPC240|MARCHF2|E3 ubiquitin-protein ligase MARCHF2|Membrane-associated RING finger protein 2|Membrane-associated RING-CH protein II|RING finger protein 172|RING-type E3 ubiquitin transferase MARCHF2|MARCH-IIProtein/peptide typeRecombinantSpecieHumanSorgente luminosaE. coliConiugazioneNon coniugatoSenza animaliYAttività biologicaStandardPurezza90%FormaLyophilisedFormulazionepH: 7,4 - 8, Constituents: 6% Trehalose, 0,14% PBSCondizioni di conservazioneStore at −20 °CTemperatura di spedizione20Applicazioni testateSDS-PAGENew Product
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



