Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant ARPC2 (from E. coli)

Human Recombinant ARPC2 (from E. coli)

Codice catalogo ABCAAB140555-20
Marketsource
Nome del fornitore:   Abcam
Human Recombinant ARPC2 (from E. coli)
Codice catalogo ABCAAB140555-20
Nome del fornitore:  Abcam
Marketsource
Numero CAS:  
Human Recombinant ARPC2 (from E. coli)
Human Recombinant ARPC2 (from E. coli)
Numero CAS:  
Price
519.00 SFr.
Log in per disponibilità
1 * 20 µG
Product Details & Documents

Recombinant Human ARPC2 protein is a Human Full Length protein, in the 1 to 300 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Senza animali:
    N
  • Protein/peptide name:
    ARPC2
  • Purezza:
    <gt/>90%
  • Attività biologica:
    Standard
  • Protein synonyms:
    ARC34|PRO2446|ARPC2|Actin-related protein 2/3 complex subunit 2|Arp2/3 complex 34 kDa subunit|p34-ARC
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Sorgente luminosa:
    E. coli
  • Cat. No.:
    ABCAAB140555-20
  • Condizioni di conservazione:
    Store at −20 °C
  • Protein/peptide type:
    Recombinant
  • Forma:
    Liquid
  • Applicazioni testate:
    MS, SDS-PAGE
  • Formulazione:
    pH: 8, Constituents: 50% Glycerol (glycerin, glycerine), 0,88% Sodium chloride, 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Temperatura di spedizione:
    20
  • Concentrazione:
    0,25 mg/ml

Specifiche