Human Lambda Light chain Antibody Pair - BSA and Azide free
Codice catalogo ABCAAB308547-10X96
Nome del fornitore: Abcam
Numero CAS:
Specifiche
- Descrizione:Human Lambda Light chain Antibody Pair - BSA and Azide free
- Test per kit:10×96 tests
Specifiche
Informazioni su questo articolo
Human Lambda Light chain Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Lambda Light chain.