Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human BMPER ELISA Kit

Human BMPER ELISA Kit

Human BMPER ELISA Kit, 96 tests
Catalog # ABCAAB309412-96TST
New Product
Supplier:   Abcam
Human BMPER ELISA Kit
Catalog # ABCAAB309412-96TST
Supplier:  Abcam
New Product
CAS Number:  
Human BMPER ELISA Kit
Human BMPER ELISA Kit

Human BMPER ELISA Kit, 96 tests

CAS Number:  
Price
SFr. 923.00
Log in for availability
1 * 1 Tests
Product Details & Documents

Human BMPER ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human BMPER in Human in Plasma, Serum, Cell culture supernatant samples.

Specifications

Specifications

  • Descrizione:
    Human BMPER ELISA Kit
  • Test per kit:
    96 tests
  • Sensibilità:
    0,45 ng/ml
  • Cat. No.:
    ABCAAB309412-96TST
  • Tipo di test:
    Sandwich
  • Metodo di rilevamento:
    Colorimetric
  • Range di rilevamento:
    0,45 - 100 ng/ml
  • Primary antibody reactivity:
    Human
  • Target protein:
    BMPER
  • Stato regolamentazione:
    RUO
  • Durata del test:
    Multiple steps
  • Tiipo del campione:
    Plasma, Serum, Cell culture supernatant
  • Temperatura di conservazione:
    4 °C
  • Formato:
    Pre-coated

Specifications