Human Synapsin I Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253633-10X96
Anbieter: Abcam
CAS-Nummer:
Technische Daten
- Durata del test:Multiple steps
- Formato:Uncoated
- Target protein:Synapsin I
- Descrizione:Human Synapsin I antibody pair - BSA and azide free
- Range di rilevamento:0,2 - 10 ng/ml
- Temperatura di conservazione:4 °C
- Stato regolamentazione:RUO
- Test per kit:10×96 tests
Technische Daten
Über diesen Artikel
Synapsin I antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Synapsin I.