Mouse E-selectin ELISA Kit (CD62E)
Mouse E-selectin ELISA kit (CD62E), 96 tests
Codice catalogo ABCAAB171182-96
Nome del fornitore: Abcam
Mouse E-selectin ELISA Kit (CD62E)
Codice catalogo ABCAAB171182-96
Nome del fornitore: Abcam
Numero CAS:
Vai a
Product Details & Documents
Mouse E-selectin ELISA kit (CD62E) is a sandwich (quantitative) ELISA kit for the measurement of Mouse E-selectin (CD62E) in mouse in plasma, cell culture supernatant, serum samples.
Specifiche
Specifiche
- Descrizione:Mouse E-selectin ELISA kit (CD62E)
- Test per kit:96 tests
- Sensibilità:10 pg/ml
- Cat. No.:ABCAAB171182-96
- Tipo di test:Sandwich
- Metodo di rilevamento:Colorimetric
- Range di rilevamento:62,5 - 4000 pg/ml
- Primary antibody reactivity:Mouse
- Target protein:CD62E
- Stato regolamentazione:RUO
- Durata del test:Multiple steps
- Tiipo del campione:Plasma, Cell culture supernatant, Serum
- Temperatura di conservazione:4 °C
- Formato:Pre-coated
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



