Human Cleaved Caspase-3 (Asp175) Antibody Pair - BSA and Azide free
Catalog # ABCAAB243998-10X96
Supplier: Abcam
CAS Number:
Specifications
- Descrizione:Human Cleaved Caspase-3 (Asp175) Antibody Pair - BSA and Azide free
Specifications
About this item
Human Cleaved Caspase-3 (Asp175) Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Cleaved Caspase-3 (Asp175).