Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human Factor VII ELISA Kit

Human Factor VII ELISA Kit

Human Factor VII ELISA kit, 96 tests
Catalog # ABCAAB108829-96
Supplier:   Abcam
Human Factor VII ELISA Kit
Catalog # ABCAAB108829-96
Supplier:  Abcam
CAS Number:  
Human Factor VII ELISA Kit
Human Factor VII ELISA Kit

Human Factor VII ELISA kit, 96 tests

CAS Number:  
Price
SFr. 784.00
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Factor VII ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Factor VII in Human in plasma, Serum samples.

Specifications

Specifications

  • Descrizione:
    Human Factor VII ELISA kit
  • Test per kit:
    96 tests
  • Tipo di test:
    Sandwich
  • Tempo ai risultati:
    4 h 0 min
  • Range di rilevamento:
    3,125 - 200 ng/ml
  • Primary antibody reactivity:
    Human
  • Stato regolamentazione:
    RUO
  • Formato:
    Pre-coated
  • Sensibilità:
    2,3 ng/ml
  • Cat. No.:
    ABCAAB108829-96
  • Metodo di rilevamento:
    Colorimetric
  • Target protein:
    Factor VII
  • Durata del test:
    Multiple steps
  • Tiipo del campione:
    Plasma, serum
  • Temperatura di conservazione:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.