Mouse/Rat IL-17A Antibody Pair - BSA and Azide free
Catalog # ABCAAB242038-10X96
Supplier: Abcam
CAS Number:
Specifications
- Descrizione:Mouse/Rat IL-17A antibody pair - BSA and Azide free
- Test per kit:10×96 tests
Specifications
About this item
Mouse/Rat IL-17A Antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse/Rat IL-17A.