ContactUsLinkComponent
rubipor XS 2001 Assembly Gloves, 60276 Series
rubipor XS 2001 Assembly Gloves, 60276 Series
Supplier:  UVEX
Certificates
rubipor XS 2001 Assembly Gloves, 60276 Series
Supplier:  UVEX
Specifications
  • Catalog No:
  • UVEX6027607
  • UVEX6027608
  • UVEX6027609
  • UVEX6027605
  • UVEX6027610
  • UVEX6027606
  • Ambidestro/mano specifico:
  • mano specifico
  • mano specifico
  • mano specifico
  • mano specifico
  • mano specifico
  • mano specifico
  • Monouso/riutilizzabile:
  • monouso
  • monouso
  • monouso
  • monouso
  • monouso
  • monouso
  • Talcati/non talcati:
  • Non talcati
  • Non talcati
  • Non talcati
  • Non talcati
  • Non talcati
  • Non talcati
  • Norma EN:
  • EN 420:2003 + A1:2009/EN 388:2016
  • EN 420:2003 + A1:2009/EN 388:2016
  • EN 420:2003 + A1:2009/EN 388:2016
  • EN 420:2003 + A1:2009/EN 388:2016
  • EN 420:2003 + A1:2009/EN 388:2016
  • EN 420:2003 + A1:2009/EN 388:2016