unigrip 6620 Assembly Gloves, 60135 Series
: UVEX
A 13-gauge knitted gloves with grip dots for precise mechanical work.
- Palm and fingers coated with PVC dots
- Thumb joints and fingertips reinforced with larger grip dots
- Good grip in dry areas with the thin PVC dots
Safety gloves in the uvex unigrip range are distinguished above all by their excellent grip in dry working environments. Their perfect fit and low weight make them extremely comfortable to wear.
- Catalog No:
- UVEX6013501
- UVEX6013502
- UVEX6013503
- UVEX6013504
- UVEX6013505
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- monouso
- monouso
- Talcati/non talcati:
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Non talcati
- Norma EN:
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018
Frequently Bought Together