Anti-Proteasome Activator Subunit 4/PSME4 Rabbit Polyclonal Antibody
Codice catalogo ABCAAB157158-100
Nome del fornitore: Abcam
Specifiche
- Taglia:100 µg
- Tipo di conf.:Vial
Specifiche
Informazioni su questo articolo
Rabbit polyclonal to Proteasome Activator Subunit 4/PSME4.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Proteasome Activator Subunit 4/PSME4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human